Benserazide hydrochloride ≥99%
Supplier: Enzo Life Sciences
Synonyms:
DL-Serine 2-(2,3,4-Trihydroxybenzyl)hydrazide Hydrochloride
, 2-Amino-3-hydroxy-N'-(2,3,4-trihydroxybenzyl)propanehydrazide hydrochloride
BML-EI165-1000
76203-476EA
72.44
USD
76203-476
Benserazide hydrochloride ≥99%
Benserazide hydrochloride
Peripheral decarboxylase inhibitor. Also inhibits tryptophan hydroxylase and tyrosine hydroxylase. Used with L-DOPA as a Parkinson’s disease therapeutic.
- Tan solid
- Soluble in water. Almost insoluble in 100% ethanol, insoluble in acetone.
Formula:
C₁₀H₁₅N₃O₅ MW: 293.70 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078571 CAS Number: 14919-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...