2-Morpholino-8-phenylchromone

Supplier: Promega Corporation

Synonyms: 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyranLY 2940022-Morpholino-8-phenyl-4H-chromen-4-one

V1201
PAV1201EA 221.06 USD
PAV1201
2-Morpholino-8-phenylchromone
2-Morpholino-8-phenylchromone

LY 294002 is a potent and specific cell-permeable inhibitor of phosphatidylinositol 3-kinases (PI 3-kinases).


  • Potent Inhibitor of Phosphatidylinositol 3-Kinases
  • Cell-permeable
  • Specifically inhibits PI 3-kinases
  • Supplied as a dried powder


LY 294002 [2-(4-Morpholinyl)-8-phenyl-4 H-1-benzopyran-4-one] is a potent and specific cell-permeable inhibitor of phosphatidylinositol 3-kinases (PI 3-kinases) with IC50 values in the 1-50uM range. LY 294002 competitively inhibits ATP binding to the catalytic subunit of PI 3-kinases and does not inhibit PI 4-kinase, DAG-kinase, PKC, PKA, MAPK, S6 kinase, EGFR or c-src tyrosine kinases and rabbit kidney ATPase.

Formula: C₁₉H₁₇NO₃
MW: 307.35 g/mol
MDL Number: MFCD00270881
CAS Number: 154447-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR