EnduRen In Vivo Renilla Luciferase Substrate, 0.34mg, Promega
Supplier: Promega
P1112
PAP1112EA 1144.03 USD
PAP1112
EnduRen In Vivo Renilla Luciferase Substrate, 0.34mg, Promega
Cell Culture Supplements and Additives
EnduRen in vivo Renilla Luciferase Substrate is a uniquely engineered coelenterazine-based compound with protected oxidation sites. These modifications are designed to minimize substrate degradation and autoluminescence.
  • Coelenterazine Substrate for Measuring Renilla Luciferase Luminescence in Animal Studies
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings
EnduRen in vivo Renilla Luciferase Substrate is a uniquely engineered coelenterazine-based compound with protected oxidation sites. These modifications are designed to minimize substrate degradation and autoluminescence. It is reported that EnduRen Substrate may have a longer kinetic output when compared to the native coelenterazine substrate when used in an in vivo imaging application in a mouse model.