Amino acid mixture 1 mM Minus Cysteine
L4471
PAL4471EA 72.77 USD
PAL4471
Amino acid mixture 1 mM Minus Cysteine
Amino acid mixture
Amino Acid Mixture, Complete, is an aqueous solution containing 1mM each of the 20 essential amino acids.
  • 1mM Solution of 20 Essential Amino Acids or Combinations Thereof
  • Amino acid solution for use with Flexi(R), TnT(R) and Rabbit Reticulocyte Lysates, Wheat Germ Extract and E. coli S30 Systems
  • Choose from mix of 20 amino acids or special mixtures indicated
  • Supplied at a concentration 1mM
This mixture is compatible for use in the Flexi Lysate, TNT Lysate and standard Rabbit Reticulocyte Lysate Systems as well as in the Wheat Germ Extract and E. coli S30 Systems.
Amino Acid Mixtures are also available lacking cysteine, methionine and cysteine, leucine or methionine.
Storage: LESS THAN -65°C