1-Boc-piperidin-4-ylboronic acid pinacol ester ≥95%
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Piperidine-4-boronic acid, pinacol ester, N-BOC protected
, tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
19802-1G
102597-364EA
166.43
USD
102597-364
1-Boc-piperidin-4-ylboronic acid pinacol ester ≥95%
1-Boc-piperidin-4-ylboronic acid pinacol ester
Formula:
C₁₆H₃₀BNO₄ MW: 311.2247 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09997792 CAS Number: 1048970-17-7 |
Specification Test Results
Analysis by Mass Spectra | Conforms to Structure |
1H NMR | Conforms to Structure |
Purity | ≥95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...