1,2-Diphenylhydrazine ≥95%

Supplier: Pfaltz & Bauer
Danger

Synonyms: Hydrazobenzene

H08360-25G
101185-405EA 220.42 USD
101185-405
1,2-Diphenylhydrazine ≥95%
1,2-Diphenylhydrazine
Formula: C₁₂H₁₂N₂
MDL Number: MFCD00003012
CAS Number: 122-66-7
EINECS: 204-563-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Purity 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR