Creatine phosphate dipotassium salt ≥98% (by TLC), Calbiochem®
Supplier: MilliporeSigma
Synonyms:
Phosphocreatine
237911-250MG
237911-1GM
237911-250MG
80051-254EA
476.29
USD
CA80051-256
80051-254
80051-256
Creatine phosphate dipotassium salt ≥98% (by TLC), Calbiochem®
Creatine phosphate dipotassium salt
Creatine phosphate, dipotassium salt is a high-energy phosphate compound for transfer of phosphate groups in the creatine phosphate energy shuttle.
Synonym: Phosphocreatine.
Formula:
C₄H₈K₂N₃O₅P Storage Temperature: Freezer |
CAS Number:
18838-38-5 |
Specification Test Results
Purity (TLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...