4-Methoxyphenylboronic acid, (max. 0.5% H₂O) ≥98% (by HPLC), white to off-white powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: (4-Methoxyphenyl)metaboric acidDihydroxy(4-methoxyphenyl)borane4-Methoxybenzeneboronic acid

27556-100G 27556-5G 27556-25G
102585-178EA 168.08 USD
102585-178 102585-176 102570-512
4-Methoxyphenylboronic acid, (max. 0.5% H₂O) ≥98% (by HPLC), white to off-white powder
4-Methoxyphenylboronic acid

Soluble in methanol.


Synonyms: (4-Methoxyphenyl)boranediol

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 200…209 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039139
CAS Number: 5720-07-0

Order Now

Specification Test Results

1H NMR Conforms to Structure
Water Content ≤0.5%
Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR