Anti-RBP1 Mouse Monoclonal Antibody (R-PE) [clone: RBP/872]

BNCR0872-250
75975-484EA 356 USD
75975-484
Anti-RBP1 Mouse Monoclonal Antibody (R-PE) [clone: RBP/872]
Antibodies
This antibody recognizes a protein of 21 kDa-25 kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells.

Type: Primary
Antigen: RBP1
Clonality: Monoclonal
Clone: Clone RBP/872
Conjugation: R-PE
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Goat, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR