Anti-TFRC Mouse Monoclonal Antibody (R-PE) [clone: 66IG10]

BNCR0871-250
75918-042EA 356 USD
75918-042
Anti-TFRC Mouse Monoclonal Antibody (R-PE) [clone: 66IG10]
Antibodies
This antibody recognizes a ~90-95 kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190 kDa. This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Its epitope is localized in the extracellular domain of CD71. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: Clone 66IG10
Conjugation: R-PE
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR