Anti-ESR1 Mouse Monoclonal Antibody (CF543) [clone: ER503]

BNC430503-100 BNC430503-500
75924-730EA 547.79 USD
75924-730 75925-976
Anti-ESR1 Mouse Monoclonal Antibody (CF543) [clone: ER503]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®543 is an orange fluorescent dye (Ex/Em 541/560 nm) designed for the 543 nm laser line. It yields the brightest conjugates among spectrally similar dyes.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone ER503
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR