Anti-MSN Mouse Monoclonal Antibody (CF680) [clone: MSN493]

BNC800493-500 BNC800493-100
75885-936EA 494 USD
75885-936 75885-934
Anti-MSN Mouse Monoclonal Antibody (CF680) [clone: MSN493]
Antibodies
This antibody recognizes 78 kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®680 is a near-infrared fluorescent dye (Ex/Em 681/698 nm). It yields the brightest conjugates among spectrally similar dyes, and is validated in super-resolution imaging by STORM.

Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: Clone MSN493
Conjugation: CF680
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR