This antibody recognizes 78 kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®660R is a far-red fluorescent dye (Ex/Em 663/682 nm) with superior brightness and unrivaled photostability among spectrally similar dyes.
Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: Clone MSN492
Conjugation: CF660R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human