Anti-ODC1 Mouse Monoclonal Antibody (CF568) [clone: ODC1/487]

Supplier: Biotium

BNC680487-500 BNC680487-100
75966-772EA 197.58 USD
75966-772 75966-774
Anti-ODC1 Mouse Monoclonal Antibody (CF568) [clone: ODC1/487]
Antibodies
This antibody recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: Clone ODC1/487
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR