Anti-PODXL Mouse Monoclonal Antibody (HRP) [clone: 2A4]

BNCH0483-100 BNCH0483-500
75971-314EA 511.96 USD
75971-314 75971-312
Anti-PODXL Mouse Monoclonal Antibody (HRP) [clone: 2A4]
Antibodies
Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.

Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: Clone 2A4
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR