This MAb reacts with 9-O-acetylated disialosyl group linked to the glycoprotein CD60w. CDw60 is a carbohydrate antigen present on gangliosides, platelets and a subset of CD4 , CD8 peripheral T-cells, but not on B-cells, monocytes or granulocytes. CDw60 is involved in the co-stimulation of T-cells.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®405M is a blue fluorescent dye (Ex/Em 408/452 nm) with superior brightness and photostability compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Type: Primary
Antigen: CDw60
Clonality: Monoclonal
Clone: Clone UM4D4
Conjugation: CF405M
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human