LEC chemokine shows chemotactic activity for lymphocytes and monocytes but not neutrophils. Also shows potent myelo-suppressive activity, suppresses proliferation of myeloid progenitor cells. Recombinant SCYA16 shows chemotactic activity for monocytes and THP-1 monocytes, but not for resting lymphocytes and neutrophils. Induces a calcium flux in THP-1 cells that were desensitized by prior expression to RANTES. Mainly expressed in liver, also found in spleen and thymus. Highly expressed in LPS- and IFN-gamma-activated monocytes, weakly in some lymphocytes, including natural killer cells, gamma-delta T-cells, and some T-cell clones. The ability of HCC-4 to markedly improve recognition of poorly immunogenic cells suggests that it could be an effective component of antitumor vaccines.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®660R is a far-red fluorescent dye (Ex/Em 663/682 nm) with superior brightness and unrivaled photostability among spectrally similar dyes.
Type: Primary
Antigen: CCL16
Clonality: Monoclonal
Clone: Clone LEC67
Conjugation: CF660R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human