Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL16 Mouse Monoclonal Antibody (Biotin) [clone: Clone LEC66]

Supplier: Biotium
BNCB0066-100 BNCB0066-500
LEC chemokine shows chemotactic activity for lymphocytes and monocytes but not neutrophils. Also shows potent myelo-suppressive activity, suppresses proliferation of myeloid progenitor cells. Recombinant SCYA16 shows chemotactic activity for monocytes and THP-1 monocytes, but not for resting lymphocytes and neutrophils. Induces a calcium flux in THP-1 cells that were desensitized by prior expression to RANTES. Mainly expressed in liver, also found in spleen and thymus. Highly expressed in LPS- and IFN-gamma-activated monocytes, weakly in some lymphocytes, including natural killer cells, gamma-delta T-cells, and some T-cell clones. The ability of HCC-4 to markedly improve recognition of poorly immunogenic cells suggests that it could be an effective component of antitumor vaccines.

Type: Primary
Antigen: CCL16
Clonality: Monoclonal
Clone: Clone LEC66
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μL BNCB0066-100 75956-652 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

500 μL BNCB0066-500 75956-654 Each (500µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CCL16
Antigen Name LEC Chemokine
Reactivity Human
Conjugation Biotin
Clonality Monoclonal
Antibody Type Primary
Clone LEC66
Host Mouse
Isotype IgG1, kappa
ELISA Yes
ImmunoFluorescence Yes
Gene ID Unigene ID: 10458
Concentration 0.1 mg/mL
Storage Buffer PBS/0.1% BSA/0.05% azide
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping