Anti-ECM1 Mouse Monoclonal Antibody (HRP) [clone: SC05]

BNCH0050-500 BNCH0050-100
75952-310EA 494 USD
75952-310 75952-516
Anti-ECM1 Mouse Monoclonal Antibody (HRP) [clone: SC05]
Antibodies
This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiation and tumorigenesis, all processes associated with differential secretory component expression.

Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: Clone SC05
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR