DuPont™ Tychem® 5000 Coveralls with Respirator Hood and Attached Socks
Supplier: DuPont
Tychem®
C3128TTNLG0006BN C3128TTN2X00062K C3128TTN3X0006BN C3128TTN2X000600 C3128TTN3X00062K C3128TTN4X0006BN C3128TTN5X00062K C3128TTN5X000600 C3128TTNXL0006BN C3128TTN5X0006BN C3128TTNSM000600 C3128TTNMD000600 C3128TTN4X00062K C3128TTNXL00062K C3128TTNXL000600 C3128TTNMD0006BN C3128TTNSM0006BN C3128TTNMD00062K C3128TTN4X000600 C3128TTNLG000600 C3128TTN2X0006BN C3128TTNSM00062K C3128TTN3X000600 C3128TTNLG00062K
76187-150CS 844.12 USD
76187-150 76187-140 76187-152 76187-142 76187-154 76187-298 76187-320 76187-144 76187-310 76187-156 76187-300 76187-146 76187-158 76187-312 76187-148 76187-302 76187-138 76187-314 76187-304 76187-316 76187-306 76187-318 76187-308 76187-160
DuPont™ Tychem® 5000 Coveralls with Respirator Hood and Attached Socks
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Respirator hood
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls are tan.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached respirator hood features elastic around face opening. Strong zipper ensures a tough, flexible closure. Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic opening at wrists allows for a tighter fit. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777.
Ordering information: Coveralls are available with a single or a double storm flap.