DuPont™ Tychem® 9000 Coveralls with Respirator Hood and Attached Socks
Supplier: DuPont
Tychem®
BR128TYL3X000200 BR128TYL2X000200 BR128TYLXL000200 BR128TYLMD0002BN
76187-230CS 276.66 USD
76187-230 76187-198 76187-202 76187-224
DuPont™ Tychem® 9000 Coveralls with Respirator Hood and Attached Socks
Disposable Garments
DuPont™ Tychem® 9000 offers wide-ranging protection backed by extensive fabric permeation barrier testing.
  • Respirator fit hood
  • Elastic wrists and ankles
  • Attached socks with outer boot flaps
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 9000 fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents. Tychem® 9000 offers tear, puncture and abrasion resistant fabrics for lasting protection. Tychem® 9000 garments are designed for enhanced comfort and ease of movement. These coveralls are high-visibility yellow.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask. Coveralls feature a strong zipper for tough, flexible closure. Longer zipper extends to chin for complete coverage of neck area. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists ensure a tighter fit. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777.