Would you like to log in again?
BBL™ Baird-Parker Agar Base Dehydrated Culture Media, BD
Supplier: BD
BBL™
297725 297214
Baird-parker agar base is used with EY (egg yolk) tellurite enrichment in the preparation of egg-tellurite-glycerine-pyruvate agar (ETGPA) for selective isolation and enumeration of coagulase-positive staphylococci from food, skin, soil, air and other materials.
It is also used for the for identification of staphylococci on the basis of their ability to clear egg yolk.
ORDER
Expand All / Collapse All
Description pH Packaging Supplier No. VWR Catalog Number Unit Price Quantity
BBL™ Baird-Parker Agar Base 6.9 ± 0.1 100 Case 297725 90004-982 Case of 100 Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
BBL™ Baird-Parker Agar Base 6.9 ± 0.1 20 Pack 297214 90008-610 Pack of 20 Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty