Ammonia solution 10% (w/w) in water, Extra Pure

Danger

Synonyms: Ammonium hydroxide

AC45868-0025 AC45868-0010
103534-382EA 32.54 USD
103534-382 103534-384
Ammonia solution 10% (w/w) in water, Extra Pure
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Density: 0.9 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Strength 9.5 to 10.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR