2-(Trifluoromethyl)pyrrolidine 97%

Warning

Synonyms: 2-(Trifluoromethyl)tetrahydropyrrole1-Aza-2-(trifluoromethyl)cyclopentane

AC45481-0050 AC45481-0010
103531-200EA 179.6 USD
103531-200 103531-202
2-(Trifluoromethyl)pyrrolidine 97%
2-(Trifluoromethyl)pyrrolidine
Formula: C₅H₈F₃N
MW: 139.12 g/mol
Boiling Pt: 110…112 °C
Density: 1.21 g/cm³
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02663405
CAS Number: 109074-67-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to orange to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR