Isohexane (mixture of isomeric branched chain Hexanes) contains <5% n-Hexane for HPLC

Danger

Synonyms: Hexanes

AC38381-0010 AC38381-0025
103530-298EA 503.08 USD
103530-298 103530-300
Isohexane (mixture of isomeric branched chain Hexanes) contains <5% n-Hexane for HPLC
Isohexane (mixture of isomeric branched chain Hexanes)
Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 60 °C (760 mmHg)
Melting Pt: –154 °C
Density: 0.66 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 73513-42-5
UN: 1208
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.01 %
UV at 250 nm A: ≤0.01
UV at 260 nm A: ≤0.005
UV 1 cm cell vs HPLC-grade water
UV at 200 nm A: ≤0.3
UV at 210 nm A: ≤0.15
UV at 220 nm A: ≤0.07
UV at 230 nm A: ≤0.03
UV at 240 nm A: ≤0.02
Residue after evaporation ≤2 ppm
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR