3-Phenylpropyl isocyanate 97%

Danger

AC45738-0010 AC45738-0050
103534-328EA 212.23 USD
103534-328 103534-326
3-Phenylpropyl isocyanate 97%
3-Phenylpropyl isocyanate
Formula: C₁₀H₁₁NO
Boiling Pt: 115 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD04039909
CAS Number: 68664-23-3
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR