Oxamyl 95%

Danger

AC45614-0010
103534-272EA 54.3 USD
103534-272
Oxamyl 95%
Oxamyl
Formula: C₇H₁₃N₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD00055334
CAS Number: 23135-22-0
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 101 °C
Water 1 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR