(1-Methyl-1H-pyrazol-5-yl)methylamine 97%

Danger

Synonyms: 5-(Aminomethyl)-1-methyl-1H-pyrazole

AC45603-0010 AC45603-0050
103534-262EA 196 USD
103534-262 103534-264
(1-Methyl-1H-pyrazol-5-yl)methylamine 97%
(1-Methyl-1H-pyrazol-5-yl)methylamine
Formula: C₅H₉N₃
MW: 111.15 g/mol
Boiling Pt: 140 °C (8 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD03419808
CAS Number: 863548-52-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Water ≤1.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR