Methylpropyldichlorosilane 97%

Danger

Synonyms: Silane, dichloromethylpropyl-dichloro(methyl)(propyl)silaneDichloro(methyl)propylsilane

AC43288-0250
103533-362EA 141.3 USD
103533-362
Methylpropyldichlorosilane 97%
Methylpropyldichlorosilane
Formula: C₄H₁₀Cl₂Si
MW: 157.109 g/mol
Boiling Pt: 125 °C (760 mmHg)
Density: 1.046 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00013608
CAS Number: 4518-94-9
UN: 2986
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (may darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm (BHT)
Peroxides ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR