4-Methoxy-2-methylphenylmagnesium bromide 0.5 M in THF, AcroSeal™

Danger

AC43181-0500
103530-350EA 143.2 USD
103530-350
4-Methoxy-2-methylphenylmagnesium bromide 0.5 M in THF, AcroSeal™
4-Methoxy-2-methylphenylmagnesium bromide
Formula: C₈H₉BrMgO
Density: 0.948 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD01319918
CAS Number: 148706-30-3
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to brown Solution
Titration Grignard 0.40 to 0.60 M
Identification Positive
Bismuth (Bi) ≥80.0 % (on dry basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR