Sodium trimethylsilanolate 1 M in dichloromethane, AcroSeal™

Danger

Synonyms: Silanol, 1,1,1-trimethyl-, sodium salt (1:1)sodium trimethyl(oxido)silaneSilanol, trimethyl-, sodium salt

AC42884-8000
103529-676EA 555.2 USD
103529-676
Sodium trimethylsilanolate 1 M in dichloromethane, AcroSeal™
Sodium trimethylsilanolate
Formula: C₃H₉NaOSi
MW: 112.179 g/mol
Density: 1.28 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00013926
CAS Number: 18027-10-6
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to amber Solution (May develop some turbidity or precipitate)
Titration with HClO4 0.90 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR