(R)-(+)-2-Amino-3-benzyloxy-1-propanol 97%

Danger

Synonyms: H-Serinol(Bzl)(2R)-2-amino-3-(benzyloxy)propan-1-ol(R)-2-amino-3-benzyloxypropan-1-ol1-Propanol, 2-amino-3-(phenylmethoxy)-, (2R)-

AC39962-0010
103533-044EA 67.7 USD
103533-044
(R)-(+)-2-Amino-3-benzyloxy-1-propanol 97%
(R)-(+)-2-Amino-3-benzyloxy-1-propanol
Formula: C₁₀H₁₅NO₂
MW: 181.235 g/mol
Boiling Pt: 307 °C (760 mmHg)
Melting Pt: 34…37 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00235943
CAS Number: 58577-87-0
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation +3.5° to +4.5° (20 °C, 589 nm) (C=1, dichloromethane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR