4-Cyanophenylacetic acid 97%

Warning

Synonyms: 2-(4-Cyanophenyl)acetic acid

AC43773-0050 AC43773-0010
103530-604EA 146.46 USD
103530-604 103530-606
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06798066
CAS Number: 5462-71-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 150.0 to 155.0 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR