4-Methyl-1,3-thiazole-5-sulfonyl chloride 97%

Danger

Synonyms: 4-Methylthiazole-5-sulfonyl chloride4-Methylthiazole-5-sulphonyl chloride

AC45364-0010 AC45364-0050
103534-150EA 506 USD
103534-150 103534-148
4-Methyl-1,3-thiazole-5-sulfonyl chloride 97%
4-Methyl-1,3-thiazole-5-sulfonyl chloride
Formula: C₄H₄ClNO₂S₂
MW: 197.66 g/mol
Melting Pt: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD09864968
CAS Number: 953070-51-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow to brown Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR