1-Benzyl-3-pyrroline 97%

Danger

Synonyms: 1-Benzyl-2,5-dihydropyrrole

AC43677-0010 AC43677-0050
103529-840EA 37.1 USD
103529-840 103533-506
1-Benzyl-3-pyrroline 97%
1-Benzyl-3-pyrroline
Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 92 °C (1hPa)
Density: 0.837 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00012324
CAS Number: 6913-92-4
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR