1-Bromo-4-(1,1-difluoroethyl)benzene 95%

Danger

AC43253-0050 AC43253-0100
103533-334EA 307 USD
103533-334 103533-336
1-Bromo-4-(1,1-difluoroethyl)benzene 95%
1-Bromo-4-(1,1-difluoroethyl)benzene
Formula: C₈H₇BrF₂
Boiling Pt: 80…82 °C (10Torr)
Storage Temperature: Ambient
MDL Number: MFCD11110333
CAS Number: 1000994-95-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Density 1.76 to 1.80 g/ml (25 °C)
Additional info distillation range (between 90 °C and 107°C): ≥90%
Additional info residue: ≤10 ug/ml
Additional info pour point: ≤-62°C
Additional info dielectric strength: ≥35 kV
Viscosity 0.7 to 0.9 cSt (at 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR