2-Methoxytetrahydropyran 98%

Warning

Synonyms: 2-Methoxytetrahydro-2H-pyran

AC45083-0250 AC45083-0050
103531-138EA 169 USD
103531-138 103531-136
2-Methoxytetrahydropyran 98%
2-Methoxytetrahydropyran
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 128…129 °C (760 mmHg)
Density: 0.963 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00075264
CAS Number: 6581-66-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR