2-(4-Fluorophenyl)propan-2-amine 97%

Danger

Synonyms: alpha, alpha-Dimethyl-4-fluorobenzylamine1-(4-Fluorophenyl)-1-methylethylamine

AC43106-0010 AC43106-0050
103533-286EA 216 USD
103533-286 103533-288
2-(4-Fluorophenyl)propan-2-amine 97%
2-(4-Fluorophenyl)propan-2-amine
Formula: C₉H₁₂FN
MW: 153.2 g/mol
Boiling Pt: 40 °C (7mbar)
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07777176
CAS Number: 17797-10-3
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR