2-Isocyanato-1-methoxy-4-methylbenzene 98%

Danger

Synonyms: 2-Methoxy-5-methylphenyl isocyanate

AC44177-0050 AC44177-0010
103533-712EA 53.1 USD
103533-712 103533-710
2-Isocyanato-1-methoxy-4-methylbenzene 98%
2-Isocyanato-1-methoxy-4-methylbenzene
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 240 °C (760 mmHg)
Melting Pt: 25…26 °C
Density: 1.112 g/cm³
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00013869
CAS Number: 59741-04-7
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR