Methylsulfonylacetonitrile 97%

Danger

Synonyms: MethanesulphonylacetonitrileMethanesulfonylacetonitrileCyanomethylsulphonylmethaneCyanomethylsulfonylmethane2-(Methylsulphonyl)acetonitrile2-(Methylsulfonyl)acetonitrile

AC43842-0250 AC43842-0050
103529-846EA 87.7 USD
103529-846 103529-848
Methylsulfonylacetonitrile 97%
Methylsulfonylacetonitrile
Formula: C₃H₅NO₂S
MW: 119.14 g/mol
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00014743
CAS Number: 2274-42-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 81 to 86 °C
Elemental Analysis 29.03 to 31.45 % (C)
Elemental Analysis 11.29 to 12.23 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR