Methyl-3-cyclohexene-1-carboxylate 97%

Warning

Synonyms: Methyl 3-cyclohexenecarboxylate3-Cyclohexene-1-carboxylic Acid Methyl EsterMethyl cyclohex-3-enecarboxylate

AC45286-5000 AC45286-1000 AC45286-0250
103531-172EA 49.8 USD
103531-172 103531-174 103531-176
Methyl-3-cyclohexene-1-carboxylate 97%
Methyl-3-cyclohexene-1-carboxylate
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 185 °C
Density: 1.02 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00053266
CAS Number: 6493-77-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4620 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR