Cyclohexylhydrazine hydrochloride 98%

Danger

AC43002-0050 AC43002-0250
103529-698EA 538.46 USD
103529-698 103533-284
Cyclohexylhydrazine hydrochloride 98%
Cyclohexylhydrazine hydrochloride
Formula: C₆H₁₄N₂
Melting Pt: 110…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060160
CAS Number: 24214-73-1
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 110 to 115 °C
Elemental Analysis 46.64 to 49.03 % C
Elemental Analysis 18.13 to 19.07 % N
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR