Chloro(decyl)dimethylsilane 97%

Danger

Synonyms: Decyldimethylchlorosilane1-(Chlorodimethylsilyl)decane

AC43219-0050 AC43219-0250
103529-754EA 99 USD
103529-754 103529-752
Chloro(decyl)dimethylsilane 97%
Chloro(decyl)dimethylsilane
Formula: C₁₂H₂₇ClSi
MW: 234.88 g/mol
Boiling Pt: 98 °C (3hPa)
Density: 0.866 g/cm³
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00054168
CAS Number: 38051-57-9
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR