1-(2-Chloro-4-pyridyl)-3-phenylurea 98%

Warning

Synonyms: Forchlorfenuron

AC45629-0010 AC45629-0050 AC45629-0250
103534-282EA 330 USD
103534-282 103534-278 103534-280
1-(2-Chloro-4-pyridyl)-3-phenylurea 98%
1-(2-Chloro-4-pyridyl)-3-phenylurea
Formula: C₁₂H₁₀ClN₃O
MW: 247.68 g/mol
Melting Pt: 165…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059898
CAS Number: 68157-60-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 170 to 177 °C
Loss on drying ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR