(Triphenylphosphoranylidene)acetonitrile 96%

Warning

Synonyms: 2-(Triphenylphosphoranylidene)acetonitrile(Cyanomethylene)triphenylphosphorane

AC37157-0050
103532-664EA 125 USD
103532-664
(Triphenylphosphoranylidene)acetonitrile 96%
(Triphenylphosphoranylidene)acetonitrile
Formula: C₂₀H₁₆NP
MW: 301.33 g/mol
Melting Pt: 189…198 °C
Storage Temperature: Freezer
MDL Number: MFCD00567633
CAS Number: 16640-68-9
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 198 °C
Additional info This product unavoidably and progressively degrades
Additional info The actual assay is mentioned on the COA.
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR