(S)-2-(Aminomethyl)pyrrolidine 98%

Danger

Synonyms: (S)-(+)-2-(Aminomethyl)pyrrolidine

AC34838-0010 AC34838-2500
103530-128EA 253 USD
103530-128 103530-130
(S)-2-(Aminomethyl)pyrrolidine 98%
(S)-2-(Aminomethyl)pyrrolidine
Formula: C₅H₁₂N₂
MW: 100.17 g/mol
Boiling Pt: 65 °C (11 mmHg)
Density: 0.933 g/cm³
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191745
CAS Number: 69500-64-7
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +19.5° to +20.5° (20 °C, 589 nm) (C=1, Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR