(3S)-(-)-3-(Dimethylamino)pyrrolidine 97%

Danger

Synonyms: (S)-3-(Dimethylamino)pyrrolidine(S)-(-)-3-(Dimethylamino)pyrrolidine

AC42645-0010
103533-156EA 203 USD
103533-156
(3S)-(-)-3-(Dimethylamino)pyrrolidine 97%
(3S)-(-)-3-(Dimethylamino)pyrrolidine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 164 °C (760 mmHg)
Density: 0.899 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00191348
CAS Number: 132883-44-4
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4660 (20 °C, 589 nm)
GC 96.0 %
Specific optical rotation -12° to -16° (20 °C, 589 nm) (c=1% ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR