(3R)-(-)-1-Benzyl-3-aminopyrrolidine 99%

Danger

Synonyms: (R)-1-Benzylpyrrolidin-3-amine(R)-(-)-1-Benzyl-3-aminopyrrolidine

AC43789-0010 AC43789-0050
103533-548EA 246.9 USD
103533-548 103533-546
(3R)-(-)-1-Benzyl-3-aminopyrrolidine 99%
(3R)-(-)-1-Benzyl-3-aminopyrrolidine
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
Boiling Pt: 108…115 °C (5 mmHg)
Density: 1.02 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00082638
CAS Number: 114715-39-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation -1° to -3° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR