3-Oxetanamine 95%

Danger

Synonyms: 3-amino-oxetane3-Aminooxetane

AC45121-5000 AC45121-2500
103534-068EA 222 USD
103534-068 103534-066
3-Oxetanamine 95%
3-Oxetanamine
Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 99 °C
Flash Pt: 24 °C
Storage Temperature: Freezer
MDL Number: MFCD08544363
CAS Number: 21635-88-1
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR