1-Bromo-4-(difluoromethyl)benzene 97%

Warning

Synonyms: 4-Bromobenzal fluoride

AC43255-0050 AC43255-0010
103533-340EA 329.6 USD
103533-340 103533-338
1-Bromo-4-(difluoromethyl)benzene 97%
1-Bromo-4-(difluoromethyl)benzene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 74…76 °C (12Torr)
Storage Temperature: Ambient
MDL Number: MFCD06657964
CAS Number: 51776-71-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR